Staticmaster® Electrostatic Discharge Workstation Kit with 1U400 Ionizer, NRD
75794-890
- Description:ESD workstation kit with 1U400 ionizer
- Cat. no.:75794-890
- Supplier No.:SMPW-KITB
This Staticmaster® PW basics kit B is essential for eliminating static charge in the work space and providing in-chamber ionization for a smaller balance chamber.
- Eliminates static charge
- Provides in-chamber ionization
Includes the 5021 ESD workstation kit, (1) 1U400 ionizer, (1) BF3 EZ mount, and (1) 5060 grounding kit.