Staticmaster® EZ Mounts, NRD | Avantor
Staticmaster® EZ Mounts, NRD
Supplier:   NRD Staticmaster
Staticmaster® EZ Mounts, NRD
Supplier:  NRD Staticmaster
Certificates
Staticmaster® EZ Mounts, NRD
Staticmaster® EZ Mounts, NRD
2 Options Available Below
Product Details & Documents
An indispensable accessory for mounting a 2U500 or 1U400 Ionizing Cartridge in the balance chamber.
  • Simple to use
  • Makes installation of ionizing cartridges fast and easy
Simply remove the protective layer of the double-sided tape and affix the E-Z mount to the back wall of the chamber. Once the mount is attached, installing ionizing cartridges is fast and easy.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Staticmaster 3" EZ mount for use with 2U500 ionizing cartridge
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $110.86
    $110.86
    Each
       $110.86
    Supplier Number: BF3
  • Staticmaster 1" EZ mount for use with 1U400 ionizing cartridge
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $110.86
    $110.86
    Each
       $110.86
    Supplier Number: BF4