Staticmaster® Duostat Positioner, NRD
Staticmaster Duostat positioner
Catalog # 10183-836
Supplier: NRD Staticmaster
Supplier #: BF2
Jump to:
Product Details & Documents
The Staticmaster® BF2-1000 Duostat Positioner is the ideal tool for removing static charge from filter media and fine powders.
- Increases weighing accuracy
- Useful for filter media and fine powders
- Removes static charge prior to placement in weigh chamber
The Duostat Positioner, when combined with two (2), 2U500 Ionizing Cartridges (sold separately) removes static charge from filter media and fine powders prior to placing them into the weigh chamber. Utilizing the BF2-1000 system results in increased weighing accuracy and efficient, reliable measurement.
Specifications
- Désignation:Staticmaster Duostat positioner