Corning® Screw Capped Micro Tubes, Corning
Self Standing Microtubes, with Brown Screw Cap and O-rings, 1.5 ml, Amber, Sterile
Catalog # 75831-988
Supplier:   Corning
 :  SCT-150-SS-X-S
Corning® Screw Capped Micro Tubes, Corning
Catalog # 75831-988
Supplier:  Corning
Corning® Screw Capped Micro Tubes, Corning
Self Standing Microtubes, with Brown Screw Cap and O-rings, 1.5 ml, Amber, Sterile
Price
$1,207.00
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 4,000 (1ml)
Product Details & Documents
Screw cap microtubes serve a broad range of laboratory needs including collection, processing, centrifugation, long-term storage and packaging/shipping of restriction enzymes and other reagents.
  • Withstand up to 20000 RCF
  • Certified RNase-, DNase- and endotoxin-safe for total sample integrity
Color coded caps provide a syringe port which is easily pierced for radioisotope labelling and fraction collecting.
Spécifications
  • Sterility:
    Sterile
  • Volume:
    1.5 ml
  • Color:
    Amber
  • Désignation:
    Self Standing Microtubes, with Brown Screw Cap and O-rings