pH Indicator Papers, Duotest, Machery Nagel
Duotest indicator papers combine two different indicator zones on a single strip
The zones are separated by a hydrophobic barrier, which effectively prevents mixing of the reaction colors and increases the mechanical stability.
Frequently Bought Together