Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Spécifications
- Description:Bi-Directional Needle-less Injection Site with Male Luer Lock
- Cat. no.:89134-222
- Supplier No.:LLV
Spécifications
À propos de cet article
SAI offers a variety of Luer Valves and Swabbable Injection Caps to simplify catheter access and prevent the introduction of microbial pathogens.
- Male Luer for easy catheter or harness connection
- All parts provided ETO-exposed
These caps can be used anywhere that catheter access is needed, such as harnesses, extension lines, catheters, and Huber needles.
Needle less injection site with male Luer lock are made of polycarbonate, silicone.