About this item
Isolation of RNA from cells and soft tissues.
- DNA-free: Genomic DNA removal column and DNase I included
- NGS-ready: RNA is ready for all downstream applications including next-gen sequencing, RT-qPCR, hybridization and more
The quick-RNA kits are fast RNA isolation kits designed for the easy, reliable, and rapid isolation of DNA-free RNA from a wide range of cell and tissue samples. The procedure combines a unique buffer system with zymo-spin column technology to yield high quality total RNA (including small RNAs 17 to 200 nt) in about 10 minutes.
The procedure for these RNA isolation kits is simple and fast. Add the provided RNA lysis buffer to a sample, and then purify the RNA using the zymo-spin columns. The result is highly-concentrated, DNA-free RNA that is suitable for RT-PCR, hybridization, sequencing and more.
In addition, these RNA isolation kits can be used for the enrichment of small and large RNAs into separate fractions.
Specifications
- Method / Format:Multiwell plate
- Prep size:Mini
- Refine target molecule:Total RNA
- Sample type:Cultured cellsHuman/animal cells and tissues
- Target molecule:RNA
- Sample size:≤1×106 Cells or 5 mg Tissue
- Description:Quick-RNA™ 96 Kit
- Sample capacity:10 µg
- Yield:10 µg RNA (binding capacity)
≥ 25 µl (elution volume)