Spring Scales
À propos de cet article
Ideal for general weighing, force experiments, action-reaction demonstrations.
- Dial plates display
- Metric/ounce/newtons
Spring scale is made of ABS housing with metal face plate, pointer, spring and hook. Features large bright colored dial face, easy-to-read black graduations on a frosted aluminum background.
4 Options Available Below
Product Details & Documents
Ideal for general weighing, force experiments, action-reaction demonstrations.
- Dial plates display
- Metric/ounce/newtons
Spring scale is made of ABS housing with metal face plate, pointer, spring and hook. Features large bright colored dial face, easy-to-read black graduations on a frosted aluminum background.
Product Family Options
Pully type
Informations sur le produit
- DésignationWeighing CapacityAvailabilityPrice
- Supplier Number: 80000014
- Supplier Number: 80000015
- Supplier Number: 80000017
- Supplier Number: 80000039