Boreal C-SCOPE Compound Microscopes
A robust microscope that provides crisp and clear visual and digital results!
- Infinity optics
- Plan lead-free objectives, 4, 10, 40 and 100X oil immersion
- Rackless mechanical stage
- Custom camera packages available for Trinocular model
The new Boreal C-SCOPE models are setting a new standard in microscope performance through improvements both in optical and mechanical features. Designed to be used in educational life science, medical and a variety of biological applications, these new microscopes are built around Boreal's complete understanding of the daily demands placed on a standard educational microscope.
Student proof features together with Infinity Plan optics provide fully corrected, repeatable, quality images.
: The camera packages for microscope 470231-464 are sold separately and include everything needed to connect to the microscope.
A robust microscope that provides crisp and clear visual and digital results!
- Infinity optics
- Plan lead-free objectives, 4, 10, 40 and 100X oil immersion
- Rackless mechanical stage
- Custom camera packages available for Trinocular model
The new Boreal C-SCOPE models are setting a new standard in microscope performance through improvements both in optical and mechanical features. Designed to be used in educational life science, medical and a variety of biological applications, these new microscopes are built around Boreal's complete understanding of the daily demands placed on a standard educational microscope.
Student proof features together with Infinity Plan optics provide fully corrected, repeatable, quality images.
: The camera packages for microscope 470231-464 are sold separately and include everything needed to connect to the microscope.
- Catalog No:
- 470231-462
- 470231-464
- Objectives:
- 4×, 10×, 40×, 100×
- 4×, 10×, 40×, 100×
- Stage:
- Rackless mechanical
- Rackless mechanical
- Illumination:
- 3W LED
- 3W LED
Product Family Options
- Désignation
- : 1100100403313
Specifications
Cat. No.470231-462Objectives4×, 10×, 40×, 100×StageRackless mechanicalIllumination3W LED - : 1100100403352
Specifications
Cat. No.470231-464Objectives4×, 10×, 40×, 100×StageRackless mechanicalIllumination3W LED