Reverse Action Forceps, Walter Stern
Reverse Action Forceps, Stainless steel, 12.7 cm
Catalog # 25607-112
Supplier: Walter Stern
: 300-066
Jump to:
Product Details & Documents
Reverse action forceps made of stainless steel with sharp, smooth points
Opens with finger pressure.
Specifications
- Désignation:Reverse Action Forceps
- Longueur:12.7 cm
- Matière:Stainless steel