HiScreen Capto DEAE Columns
HiScreen™ Capto™ DEAE, PP, 90 µm, 7.7×100 mm, 4.7 ml
Supplier: Cytiva
Fournisseur #: 28926982
Jump to:
Product Details & Documents
HiScreen™ Capto™ DEAE column is a weak anion exchanger with a bed height of 10 cm making it an excellent tool for method optimization, parameter screening, robustness testing, and convenient scale-up.
- Prepacked, ready-to-use columns for convenient process development optimization
- Small bed volume gives fast results and minimal sample/buffer consumption
- Easily connected in series to achieve 20 cm bed height
- Reproducible results, scalable to BioProcess columns packed with the same media using the same linear fluid velocity
- Used in an optimal way with liquid chromatography systems such as ÄKTA™
- Flow velocity: <700 cm/h
Process fluid velocities can be applied, since the bed height gives enough residence time and the results can then serve as basis for linear process scale-up. It is prepacked with Capto™ DEAE ion exchange chromatography medium.
Spécifications
- Granulométrie:90 µm
- Capacité:4.7 ml
- Désignation:HiScreen™ Capto™ DEAE
- Column Hardware:PP
- Bed Dimensions:7.7×100 mm
- Cat. No.:CA97067-804
- Max. pressure:3 bar [0.3 MPa] (44 psi)
- Storage:4...30 °C, 20% Ethanol
- pH range:2 - 9