iLog® Temperature Loggers, Cryopak Verification Technologies
Supplier: Cryopak Verification Technologies
Certificates
About this item
The user programmable iLog data logger provides technological abilities for monitoring temperature sensitive products while supporting compliance requirements. The case is made from Polycarbonate.
- Large LCD display with multi-functional viewing
- Visual and audible alarms
- Bookmark capabilities
- Wide temperature range (high heat, ambient, refrigerated, frozen and dry ice)
- High alarm; low alarm configuration
- Two year warranty
Detachable, customizable external sensor lengths and probe styles.
Certifications: ISO9001:2008, CE, RoHS, EN 12830.
Ordering information: A temperature/humidity logger is available; contact your Avantor representative for more information.
Specifications
- Catalog No:
- 10033-860
- 97017-200
- 97017-198
- 10033-854
- 10033-852
- 97017-216
- 97017-238
- 10033-866
- 10033-864
- 10033-862
- 97017-204
- 10033-858
- 10033-856
- Sensor:
- 2 ext. sensors
- 1 int. + 1 ext. sensor
- 1 int. + 1 ext. sensor
- 2 ext. sensors
- 1 int. + 1 ext. sensor
- 2 ext. sensors
- 1 int. sensor
- 2 ext. sensors
- 2 ext. sensors
- 2 ext. sensors
- 2 ext. sensors
- 2 ext. sensors
- 2 ext. sensors
- Alarm:
- Two alarm thresholds, visual LED alarm
- Two alarm thresholds, visual LED alarm
- Two alarm thresholds, visual LED alarm
- Two alarm thresholds, visual LED alarm
- Two alarm thresholds, visual LED alarm
- Two alarm thresholds, visual LED alarm
- Two alarm thresholds, visual LED alarm
- Two alarm thresholds, visual LED alarm
- Two alarm thresholds, visual LED alarm
- Two alarm thresholds, visual LED alarm
- Two alarm thresholds, visual LED alarm
- Two alarm thresholds, visual LED alarm
- Two alarm thresholds, visual LED alarm
- Display:
- Software programmable LCD and active, alarm LED's
- Software programmable LCD and active, alarm LED's
- Software programmable LCD and active, alarm LED's
- Software programmable LCD and active, alarm LED's
- Software programmable LCD and active, alarm LED's
- Software programmable LCD and active, alarm LED's
- Software programmable LCD and active, alarm LED's
- Software programmable LCD and active, alarm LED's
- Software programmable LCD and active, alarm LED's
- Software programmable LCD and active, alarm LED's
- Software programmable LCD and active, alarm LED's
- Software programmable LCD and active, alarm LED's
- Software programmable LCD and active, alarm LED's
- Software:
- Console Pro
- Console Pro
- Console Pro
- Console Pro
- Console Pro
- Console Pro
- Console Pro
- Console Pro
- Console Pro
- Console Pro
- Console Pro
- Console Pro
- Console Pro
- Protection class:
- IP 67 int. sensor; IP 65 ext. sensor
- IP 67 int. sensor; IP 65 ext. sensor
- IP 67 int. sensor; IP 65 ext. sensor
- IP 67 int. sensor; IP 65 ext. sensor
- IP 67 int. sensor; IP 65 ext. sensor
- IP 67 int. sensor; IP 65 ext. sensor
- IP 67 int. sensor; IP 65 ext. sensor
- IP 67 int. sensor; IP 65 ext. sensor
- IP 67 int. sensor; IP 65 ext. sensor
- IP 67 int. sensor; IP 65 ext. sensor
- IP 67 int. sensor; IP 65 ext. sensor
- IP 67 int. sensor; IP 65 ext. sensor
- IP 67 int. sensor; IP 65 ext. sensor
- Battery life:
- 1 - 2 years
- 1 - 2 years
- 1 - 2 years
- 1 - 2 years
- 1 - 2 years
- 1 - 2 years
- 1 - 2 years
- 1 - 2 years
- 1 - 2 years
- 1 - 2 years
- 1 - 2 years
- 1 - 2 years
- 1 - 2 years
- Response time:
- T90 of 12 min in moving air (int. sensor; T90 of 5 min (ext. sensor)
- T90 of 12 min in moving air (int. sensor; T90 of 5 min (ext. sensor)
- T90 of 12 min in moving air (int. sensor; T90 of 5 min (ext. sensor)
- T90 of 12 min in moving air (int. sensor; T90 of 5 min (ext. sensor)
- T90 of 12 min in moving air (int. sensor; T90 of 5 min (ext. sensor)
- T90 of 12 min in moving air (int. sensor; T90 of 5 min (ext. sensor)
- T90 of 12 min in moving air (int. sensor; T90 of 5 min (ext. sensor)
- T90 of 12 min in moving air (int. sensor; T90 of 5 min (ext. sensor)
- T90 of 12 min in moving air (int. sensor; T90 of 5 min (ext. sensor)
- T90 of 12 min in moving air (int. sensor; T90 of 5 min (ext. sensor)
- T90 of 12 min in moving air (int. sensor; T90 of 5 min (ext. sensor)
- T90 of 12 min in moving air (int. sensor; T90 of 5 min (ext. sensor)
- T90 of 12 min in moving air (int. sensor; T90 of 5 min (ext. sensor)
- Weight:
- 70 g
- 70 g
- 70 g
- 70 g
- 70 g
- 70 g
- 70 g
- 70 g
- 70 g
- 70 g
- 70 g
- 70 g
- 70 g
- W×D×H:
- Ø×H: 76×36 mm
- Ø×H: 76×36 mm
- Ø×H: 76×36 mm
- Ø×H: 76×36 mm
- Ø×H: 76×36 mm
- Ø×H: 76×36 mm
- Ø×H: 76×36 mm
- Ø×H: 76×36 mm
- Ø×H: 76×36 mm
- Ø×H: 76×36 mm
- Ø×H: 76×36 mm
- Ø×H: 76×36 mm
- Ø×H: 76×36 mm
- Data memory - No. of readings:
- 32000
- 32000
- 32000
- 32000
- 32000
- 32000
- 32000
- 32000
- 32000
- 32000
- 32000
- 32000
- 32000
- Batteries:
- User replaceable, 3.6 V ½ AA lithium battery
- User replaceable, 3.6 V ½ AA lithium battery
- User replaceable, 3.6 V ½ AA lithium battery
- User replaceable, 3.6 V ½ AA lithium battery
- User replaceable, 3.6 V ½ AA lithium battery
- User replaceable, 3.6 V ½ AA lithium battery
- User replaceable, 3.6 V ½ AA lithium battery
- User replaceable, 3.6 V ½ AA lithium battery
- User replaceable, 3.6 V ½ AA lithium battery
- User replaceable, 3.6 V ½ AA lithium battery
- User replaceable, 3.6 V ½ AA lithium battery
- User replaceable, 3.6 V ½ AA lithium battery
- User replaceable, 3.6 V ½ AA lithium battery