À propos de cet article
YMC-Triart C18, 10, 15 and 20 µm semi-preparative and preparative columns are packed with a novel, organic/inorganic hybrid silica particles derivatized with C18.
- Same selectivity as analytical Triart C18 particle sizes
- Chemically and mechanically stable
- Base-washable (clean in place)
- Particle sizes of 10, 15, and 20 µm
- Ability to operate in 100% aqueous environments
- High loading capacity
The core material for all of the YMC-Triart products, the particle, is inspired by YMC’s flow chemistry technology. Inherent in YMC’s process is a high level of temperature and particle formation control. As a result, Triart particles are consistent, reproducible, and scalable across many particle sizes. The hybrid Triart particles produce a material that is highly resilient and robust when used at extreme pH and elevated temperature. These particles are very stable under mechanical stress such as high pressure. YMC-Triart C18 Semi-preparative and Preparative columns can be used at temperatures up to 70 °C at pH 1 to 7, and 50 °C from pH 7 to 12.
Triart C18 columns feature chemical stability and mechanical strength, operation over a wide pH range of 1 to 12, high loading capacity, narrow particle and pore size distributions, and the ability to operate in 100% aqueous environments. Triart C18 semi-preparative and preparative columns are available in many column geometries.