VWR® Basic Industrial SPP Bouffants | Avantor
VWR® Basic Industrial SPP Bouffants
VWR® Basic Industrial SPP Bouffants
Supplier:  VWR International
Certificates
VWR® Basic Industrial SPP Bouffants
30 Options Available Below
Product Details & Documents
VWR® Basic SPP Bouffant Caps provide lightweight, breathable head coverage for controlled environments. Made from durable SPP, these caps are designed to minimize contamination by keeping hair securely contained. The non-linting material helps maintain cleanroom integrity, while the elastic closure ensures a comfortable, secure fit for various head sizes.
  • Durable SPP material provides long-lasting performance
  • Lightweight and breathable SPP ensures comfort during extended wear
  • Non-linting design maintains cleanroom and controlled environment integrity
  • Elastic closure provides a secure and comfortable fit for various head sizes
  • Latex-free construction is safe for users with latex sensitivities
  • Cost-effective disposable solution; eliminates laundering and reduces risk of cross-contamination
  • Lot-controlled production ensures consistency and compliance
VWR® Basic SPP Bouffant Caps are designed to provide reliable head coverage in cleanrooms, laboratories, and other controlled environments where contamination control is essential. Made from durable SPP, these caps offer a lightweight and breathable solution that ensures comfort during extended wear. The non-linting material helps maintain the integrity of sensitive environments by minimizing particle generation, making them ideal for applications requiring strict cleanliness standards.
Each cap features an elastic closure that provides a secure and comfortable fit for various head sizes, ensuring convenience and ease of use. Latex-free construction makes these bouffant caps suitable for individuals with latex sensitivities, while their disposable design offers a cost-effective alternative to reusable headwear, reducing laundering requirements and contamination risks. Manufactured in ISO-certified facilities and lot-controlled for consistent quality, VWR® Basic SPP Bouffant Caps deliver dependable performance for contamination-sensitive operations.
Packaging: Bouffant caps are packaged 1000 per case.
Documents
Specifications for Product Family
Spécifications
  • Catalog No:
  • 89107-768
  • 89107-770
  • 89131-620
  • 89131-622
  • 89186-064
  • 76003-494
  • 89522-670
  • 89522-672
  • 76008-534
  • 76008-536
  • 76008-538
  • 76008-540
  • 76008-542
  • 76008-544
  • 76008-546
  • 76008-548
  • 76008-550
  • 76008-552
  • 76008-514
  • 76008-516
  • 76008-518
  • 76008-520
  • 76008-522
  • 76008-524
  • 76008-526
  • 76008-528
  • 76008-530
  • 76008-532
  • 76218-552
  • 76218-554
  • Fabric:
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Cleanroom class:
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Garment application:
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Cleanroom|Industrial|Laboratory|Food
  • Particle shedding (Helmke Drum):
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Water resistance:
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • Weight:
  • 10 g/m²
  • 10 g/m²
  • 10 g/m²
  • 10 g/m²
  • 10 g/m²
  • 10 g/m²
  • 11 g/m²
  • 11 g/m²
  • 9 g/m²
  • 9 g/m²
  • 9 g/m²
  • 9 g/m²
  • 9 g/m²
  • 9 g/m²
  • 9 g/m²
  • 9 g/m²
  • 9 g/m²
  • 9 g/m²
  • 9 g/m²
  • 9 g/m²
  • 9 g/m²
  • 9 g/m²
  • 9 g/m²
  • 9 g/m²
  • 9 g/m²
  • 9 g/m²
  • 9 g/m²
  • 9 g/m²
  • 14 g/m²
  • 14 g/m²
Spécifications
Product Family Options
Basic Protection 10G SPP bouffant
Informations sur le produit
  • Color
    Ø
    Availability
    Price
  • White
    21"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $230.54
    $230.54
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 32000
    Pallet of 32000 - $7,377.15
    $7,377.15
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $230.54
    Pallet of 32000
       $7,377.15
    Spécifications:
    More
    Specifications
    Cat. No.
    89107-768
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    10 g/m²
  • White
    24"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $257.76
    $257.76
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 32000
    Pallet of 32000 - $8,248.32
    $8,248.32
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $257.76
    Pallet of 32000
       $8,248.32
    Spécifications:
    More
    Specifications
    Cat. No.
    89107-770
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    10 g/m²
  • Blue
    21"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $230.28
    $230.28
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 36000
    Pallet of 36000 - $8,290.08
    $8,290.08
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $230.28
    Pallet of 36000
       $8,290.08
    Spécifications:
    More
    Specifications
    Cat. No.
    89131-620
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    10 g/m²
  • Blue
    24"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $257.37
    $257.37
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 32000
    Pallet of 32000 - $8,235.78
    $8,235.78
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $257.37
    Pallet of 32000
       $8,235.78
    Spécifications:
    More
    Specifications
    Cat. No.
    89131-622
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    10 g/m²
  • White
    28"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $275.27
    $275.27
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 18000
    Pallet of 18000 - $4,954.90
    $4,954.90
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $275.27
    Pallet of 18000
       $4,954.90
    Spécifications:
    More
    Specifications
    Cat. No.
    89186-064
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    10 g/m²
  • Blue
    28"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $276.79
    $276.79
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $276.79
    Spécifications:
    More
    Specifications
    Cat. No.
    76003-494
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    10 g/m²
Basic Protection 11G SPP bouffant
Informations sur le produit
  • Color
    Ø
    Availability
    Price
  • White
    21"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $185.42
    $185.42
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Each
    Each - $185.42
    $185.42
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $185.42
    Each
       $185.42
    Spécifications:
    More
    Specifications
    Cat. No.
    89522-670
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    11 g/m²
  • White
    24"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $204.85
    $204.85
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Each
    Each - $204.85
    $204.85
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $204.85
    Each
       $204.85
    Spécifications:
    More
    Specifications
    Cat. No.
    89522-672
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    11 g/m²
Basic Industrial SPP bouffant
Informations sur le produit
  • Color
    Ø
    Availability
    Price
  • White
    21"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $69.30
    $69.30
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $69.30
    Spécifications:
    More
    Specifications
    Cat. No.
    76008-534
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    9 g/m²
  • White
    24"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $94.17
    $94.17
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $94.17
    Spécifications:
    More
    Specifications
    Cat. No.
    76008-536
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    9 g/m²
  • Blue
    21"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $69.35
    $69.35
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $69.35
    Spécifications:
    More
    Specifications
    Cat. No.
    76008-538
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    9 g/m²
  • Blue
    24"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $94.17
    $94.17
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $94.17
    Spécifications:
    More
    Specifications
    Cat. No.
    76008-540
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    9 g/m²
  • Red
    21"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $69.01
    $69.01
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $69.01
    Spécifications:
    More
    Specifications
    Cat. No.
    76008-542
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    9 g/m²
  • Red
    24"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $91.17
    $91.17
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $91.17
    Spécifications:
    More
    Specifications
    Cat. No.
    76008-544
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    9 g/m²
  • Yellow
    21"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $69.01
    $69.01
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $69.01
    Spécifications:
    More
    Specifications
    Cat. No.
    76008-546
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    9 g/m²
  • Yellow
    24"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $91.17
    $91.17
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $91.17
    Spécifications:
    More
    Specifications
    Cat. No.
    76008-548
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    9 g/m²
  • Green
    21"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $70.36
    $70.36
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $70.36
    Spécifications:
    More
    Specifications
    Cat. No.
    76008-550
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    9 g/m²
  • Green
    24"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $94.23
    $94.23
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $94.23
    Spécifications:
    More
    Specifications
    Cat. No.
    76008-552
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    9 g/m²
Basic Industrial pleated SPP bouffant
Informations sur le produit
  • Color
    Ø
    Availability
    Price
  • White
    21"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $51.91
    $51.91
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $51.91
    Spécifications:
    More
    Specifications
    Cat. No.
    76008-514
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    9 g/m²
  • White
    24"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $68.57
    $68.57
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $68.57
    Spécifications:
    More
    Specifications
    Cat. No.
    76008-516
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    9 g/m²
  • Blue
    21"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $55.78
    $55.78
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $55.78
    Spécifications:
    More
    Specifications
    Cat. No.
    76008-518
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    9 g/m²
  • Blue
    24"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $66.89
    $66.89
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $66.89
    Spécifications:
    More
    Specifications
    Cat. No.
    76008-520
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    9 g/m²
  • Red
    21"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $52.97
    $52.97
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $52.97
    Spécifications:
    More
    Specifications
    Cat. No.
    76008-522
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    9 g/m²
  • Red
    24"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $67.95
    $67.95
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $67.95
    Spécifications:
    More
    Specifications
    Cat. No.
    76008-524
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    9 g/m²
  • Yellow
    21"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $55.78
    $55.78
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $55.78
    Spécifications:
    More
    Specifications
    Cat. No.
    76008-526
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    9 g/m²
  • Yellow
    24"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $66.89
    $66.89
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $66.89
    Spécifications:
    More
    Specifications
    Cat. No.
    76008-528
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    9 g/m²
  • Green
    21"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $51.91
    $51.91
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $51.91
    Spécifications:
    More
    Specifications
    Cat. No.
    76008-530
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    9 g/m²
  • Green
    24"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $66.89
    $66.89
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $66.89
    Spécifications:
    More
    Specifications
    Cat. No.
    76008-532
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    9 g/m²
Basic Protection 14G SPP bouffant
Informations sur le produit
  • Color
    Ø
    Availability
    Price
  • White
    21"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $279.16
    $279.16
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $279.16
    Spécifications:
    More
    Specifications
    Cat. No.
    76218-552
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    14 g/m²
  • White
    24"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $344.26
    $344.26
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $344.26
    Spécifications:
    More
    Specifications
    Cat. No.
    76218-554
    Fabric
    Spunbound polypropylene
    Cleanroom class
    10000/ISO 7
    Gender
    Unisex
    Garment application
    Cleanroom|Industrial|Laboratory|Food
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    14 g/m²