VWR® Basic Industrial Coveralls
VWR® Basic Industrial Coveralls
VWR® Basic Industrial Coveralls
VWR® Basic Industrial Coveralls
Disposable coverall made from 20 gram spunbond polypropylene (SPP) fabric. Provides lightweight, breathable protection for nonhazardous environments. Ideal for general industrial use, food processing, and clean areas requiring basic contamination control.
  • Lightweight and breathable fabric reduces heat buildup during extended use
  • Zipper front closure for easy donning and removal
  • Elastic wrists and ankles for better coverage
  • Single-use, disposable garment for contamination control
  • Economical solution for basic protection needs
  • Non-linting material helps maintain clean environments
  • Suitable for general industrial, food processing, and nonhazardous environments
The VWR® basic protection coverall is designed to provide reliable, economical protection in environments where contamination control is essential, but exposure risks are minimal. Constructed from 20 gram spunbond polypropylene (SPP), this coverall offers a lightweight and breathable solution that ensures user comfort during extended wear. The non-woven SPP fabric is non-linting, reducing the risk of particle contamination, and is suitable for clean areas, food processing, and general industrial applications.
Key design features include a zipper front closure for easy donning and removal, along with elastic wrists and ankles to enhance coverage and minimize gaps. The disposable nature of the garment supports hygiene compliance.
 : Coveralls are packaged 25 per case.
  • Catalog No:
  • 75874-030
  • 75874-032
  • 75874-034
  • 75874-036
  • 75874-038
  • 75874-040
  • 75874-042
  • 75874-044
  • 75874-254
  • 75874-256
  • 75874-258
  • 75874-260
  • 75874-262
  • 75874-264
  • 75874-266
  • 75874-268
  • 75874-270
  • 75874-272
  • 75874-274
  • 75874-276
  • 75874-278
  • 75874-280
  • 75874-282
  • 75874-284
  • 75874-286
  • 75874-288
  • 75874-290
  • 75874-292
  • Fabric:
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Closure type:
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Cleanroom class:
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • Pockets:
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Garment application:
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Particle shedding (Helmke Drum):
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Water resistance:
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • Weight:
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
  • 20 g/m²
Product Family Options
White
  • Color
    Taille de confection
    Longueur
    Chest Size
  • White
    S
    66" (body), 28" (leg)
    25"
    Case of 25
    Case of 25 - $125.18
    $125.18
    Case of 25
       $125.18
    Specifications
    Cat. No.
    75874-030
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • White
    M
    67" (body), 29" (leg)
    26"
    Case of 25
    Case of 25 - $127.00
    $127.00
    Case of 25
       $127.00
    Specifications
    Cat. No.
    75874-032
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • White
    L
    68" (body), 30" (leg)
    27"
    Case of 25
    Case of 25 - $127.00
    $127.00
    Case of 25
       $127.00
    Specifications
    Cat. No.
    75874-034
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • White
    XL
    69" (body), 30" (leg)
    28"
    Case of 25
    Case of 25 - $127.00
    $127.00
    Case of 25
       $127.00
    Specifications
    Cat. No.
    75874-036
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • White
    XXL
    70" (body), 31½" (leg)
    29"
    Case of 25
    Case of 25 - $127.00
    $127.00
    Case of 25
       $127.00
    Specifications
    Cat. No.
    75874-038
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • White
    3XL
    71" (body), 32½" (leg)
    30"
    Case of 25
    Case of 25 - $127.84
    $127.84
    Case of 25
       $127.84
    Specifications
    Cat. No.
    75874-040
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • White
    4XL
    72" (body), 33" (leg)
    31"
    Case of 25
    Case of 25 - $22.36
    $22.36
    Case of 25
       $22.36
    Specifications
    Cat. No.
    75874-042
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
Dark blue
  • Color
    Taille de confection
    Longueur
    Chest Size
  • Dark blue
    S
    66" (body), 28" (leg)
    25"
    Case of 25
    Case of 25 - $31.96
    $31.96
    Case of 25
       $31.96
    Specifications
    Cat. No.
    75874-044
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Dark blue
    M
    67" (body), 29" (leg)
    26"
    Case of 25
    Case of 25 - $127.00
    $127.00
    Case of 25
       $127.00
    Specifications
    Cat. No.
    75874-254
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Dark blue
    L
    68" (body), 30" (leg)
    27"
    Case of 25
    Case of 25 - $127.00
    $127.00
    Case of 25
       $127.00
    Specifications
    Cat. No.
    75874-256
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Dark blue
    XL
    69" (body), 30" (leg)
    28"
    Case of 25
    Case of 25 - $127.00
    $127.00
    Case of 25
       $127.00
    Specifications
    Cat. No.
    75874-258
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Dark blue
    XXL
    70" (body), 31½" (leg)
    29"
    Case of 25
    Case of 25 - $127.00
    $127.00
    Case of 25
       $127.00
    Specifications
    Cat. No.
    75874-260
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Dark blue
    3XL
    71" (body), 32½" (leg)
    30"
    Case of 25
    Case of 25 - $128.25
    $128.25
    Case of 25
       $128.25
    Specifications
    Cat. No.
    75874-262
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Dark blue
    4XL
    72" (body), 33" (leg)
    31"
    Case of 25
    Case of 25 - $31.96
    $31.96
    Case of 25
       $31.96
    Specifications
    Cat. No.
    75874-264
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
Green
  • Color
    Taille de confection
    Longueur
    Chest Size
  • Green
    S
    66" (body), 28" (leg)
    25"
    Case of 25
    Case of 25 - $31.96
    $31.96
    Case of 25
       $31.96
    Specifications
    Cat. No.
    75874-266
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Green
    M
    67" (body), 29" (leg)
    26"
    Case of 25
    Case of 25 - $125.18
    $125.18
    Case of 25
       $125.18
    Specifications
    Cat. No.
    75874-268
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Green
    L
    68" (body), 30" (leg)
    27"
    Case of 25
    Case of 25 - $31.96
    $31.96
    Case of 25
       $31.96
    Specifications
    Cat. No.
    75874-270
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Green
    XL
    69" (body), 30" (leg)
    28"
    Case of 25
    Case of 25 - $31.96
    $31.96
    Case of 25
       $31.96
    Specifications
    Cat. No.
    75874-272
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Green
    XXL
    70" (body), 31½" (leg)
    29"
    Case of 25
    Case of 25 - $125.59
    $125.59
    Case of 25
       $125.59
    Specifications
    Cat. No.
    75874-274
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Green
    3XL
    71" (body), 32½" (leg)
    30"
    Case of 25
    Case of 25 - $14.90
    $14.90
    Case of 25
       $14.90
    Specifications
    Cat. No.
    75874-276
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Green
    4XL
    72" (body), 33" (leg)
    31"
    Case of 25
    Case of 25 - $127.77
    $127.77
    Case of 25
       $127.77
    Specifications
    Cat. No.
    75874-278
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
Red
  • Color
    Taille de confection
    Longueur
    Chest Size
  • Red
    S
    66" (body), 28" (leg)
    25"
    Case of 25
    Case of 25 - $125.18
    $125.18
    Case of 25
       $125.18
    Specifications
    Cat. No.
    75874-280
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Red
    M
    67" (body), 29" (leg)
    26"
    Case of 25
    Case of 25 - $22.78
    $22.78
    Case of 25
       $22.78
    Specifications
    Cat. No.
    75874-282
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Red
    L
    68" (body), 30" (leg)
    27"
    Case of 25
    Case of 25 - $15.06
    $15.06
    Case of 25
       $15.06
    Specifications
    Cat. No.
    75874-284
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Red
    XL
    69" (body), 30" (leg)
    28"
    Case of 25
    Case of 25 - $128.37
    $128.37
    Case of 25
       $128.37
    Specifications
    Cat. No.
    75874-286
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Red
    XXL
    70" (body), 31½" (leg)
    29"
    Case of 25
    Case of 25 - $129.40
    $129.40
    Case of 25
       $129.40
    Specifications
    Cat. No.
    75874-288
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Red
    3XL
    71" (body), 32½" (leg)
    30"
    Case of 25
    Case of 25 - $127.00
    $127.00
    Case of 25
       $127.00
    Specifications
    Cat. No.
    75874-290
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²
  • Red
    4XL
    72" (body), 33" (leg)
    31"
    Case of 25
    Case of 25 - $125.18
    $125.18
    Case of 25
       $125.18
    Specifications
    Cat. No.
    75874-292
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    20 g/m²