ContactUsLinkComponent
Radiometer Economy
Radiometer Economy
Catalog # 470230-174
Supplier:  Wards
undefined
Radiometer Economy
Catalog # 470230-174
Supplier:  Wards
Supplier Number:  PH0418A
Specifications
  • Description:
    Radiometer Economy
  • Cat. no.:
    470230-174
Specifications
About this item
Radiometer with four lightweight metal arms.
  • Includes black and white vanes in color
  • Mounted on round plastic molded base
  • Glass bulb with 70 mm dia.
Comprising partly evacuated glass bulb approx. 70 mm diameter, containing at its center a fine pivot which supports four light weight metal arms. One side of each vane is blackened, the reverse side is bright. Mounted on round plastic molded base.