Dynamics Cart, Set 3 Wheeled
Catalogue # 470230-150
Fournisseur: Wards
Spécifications
- Description:Dynamics Cart, Set 3 wheeled
- Dimensions:300 x 90 mm
- Cat. no.:470230-150
Spécifications
À propos de cet article
Low friction cart for kinematics.
- Carts have with low-friction ball-bearing wheels
This pair of cart is constructed from solid wood and aluminum. Fitted with high quality frictionless nylon ball bearing wheels allows for more accurate dynamic measurements. One of the carts is equipped with an exploder piston that is activated by a trigger on the top of the cart.