ContactUsLinkComponent
Parallelogram of Forces Apparatus
  470230-138
 :  Wards
undefined
Parallelogram of Forces Apparatus
  470230-138
 :  Wards
 :  PH0346
  • Description:
    Parallelogram of Forces Apparatus
  • Dimensions:
    25.5 x 19.5 x 0.5" (board)
  • Cat. no.:
    470230-138
Apparatus to demonstrate parallelogram of forces with accessories.
  • Slotted weights 50g each (12 total)
This apparatus is an ideal tool for studying the relationship between forces in a visually and kinesthetically effective manner.
 : Includes board, 2 pulleys on clamps, iron nickel plated hangers, and slotted weights.