ContactUsLinkComponent
Inclined Plane Steel with Pulley
Catalogue # 470230-124
Fournisseur:  Wards
undefined
Inclined Plane Steel with Pulley
Catalogue # 470230-124
Fournisseur:  Wards
Supplier Number:  PH0335
Spécifications
  • Description:
    Inclined Plane, Steel with Pulley
  • Cat. no.:
    470230-124
Spécifications
À propos de cet article
Steel inclined plane with accessories.
  • 48” long and straight as an arrow.
  • Made of heavy steel with rounded corners
  • Includes car, clamp pulley, string and basket
  • Stand base, rod and support clamp are NOT included
This green inclined plane is 48" long and straight as an arrow. It is made of heavy steel with rounded corners and can accommodate any kind of car. Holes in the supporting flanges fit a 12 mm support rod. Includes car ,clamp pulley, string and basket. Stand base, rod and support clamp are not included.