ContactUsLinkComponent
Pulley with Universal Clamp
Catalog # 470230-106
Supplier:  Wards
undefined
Pulley with Universal Clamp
Catalog # 470230-106
Supplier:  Wards
Supplier Number:  PH0299
Specifications
  • Description:
    Pulley with Universal Clamp
  • Cat. no.:
    470230-106
Specifications
About this item
Pulley with universal clamp for variety of surfaces.
  • Pulley Length 5"
  • Universal clamp allows for attachment to a variety of surfaces
The medium pulley with universal clamp is an extremely useful tool that can be positioned on a variety of surfaces, due to the universality of the clamp. This pulley provides an ideal tool for studying various concepts in physical mechanics.