Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Single Pulley Bench Mount

Single Pulley Bench Mount

Single Pulley, Bench Mount
Catalog # 470230-096
Supplier:   Wards
Supplier #:  PH0290
Single Pulley Bench Mount
Catalog # 470230-096
Supplier:  Wards
Single Pulley Bench Mount
Single Pulley Bench Mount

Single Pulley, Bench Mount

Price
$25.30
Log in pour la disponibilité
Each
Product Details & Documents

Bench mounted aluminum pulley.

  • Single pulley
  • Bench mounted
  • Aluminum
  • 50 mm dia.
  • For clamping to benches/boards up to 28 mm thick

Bench mounting, aluminum 50 mm diameter, in metal frame for clamping to benches or boards up to 28 mm thick.

Spécifications
  • Désignation:
    Single Pulley, Bench Mount
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.