ContactUsLinkComponent
Differential Pulley Economy
Catalogue # 470230-094
Fournisseur:  Wards
undefined
Differential Pulley Economy
Catalogue # 470230-094
Fournisseur:  Wards
Supplier Number:  PH0283
Spécifications
  • Description:
    Differential Pulley, Economy
  • Cat. no.:
    470230-094
Spécifications
À propos de cet article
Economical pulley for simple machine experiments.
  • One piece plastic pulley
  • Economically friendly pricing
  • 20, 40, 60 mm dia. steps
One piece, plastic differential pulley in three steps of 20, 40, 60 mm diameter, mounted with rod.