Differential Pulley Economy
Catalogue # 470230-094
Fournisseur: Wards
Spécifications
- Description:Differential Pulley, Economy
- Cat. no.:470230-094
Spécifications
À propos de cet article
Economical pulley for simple machine experiments.
- One piece plastic pulley
- Economically friendly pricing
- 20, 40, 60 mm dia. steps
One piece, plastic differential pulley in three steps of 20, 40, 60 mm diameter, mounted with rod.