Decade Resistance Box 4
Decade Resistance Box, 4
Catalog # 470230-406
Supplier: Wards
Fournisseur #: PH1133PLN10
Jump to:
Product Details & Documents
Standard decade resistance box.
- Standard decade resistance box
- Up to a total of 11,110 Ohms
- Resolution at 1, 10, 100, and 1000 Ohms
- 0.5 watt resistors
This decade resistance substitution box provides 4 decade ranges as given below and total value 0 to 11110 Ohms with a resolution of 1 Ohm. Each decade is selected using rotary switch. All resistance values are of ±1% tolerance and with 0.5 watt resistors, 0.5555 watts is the theoretical maximum.
Spécifications
- Désignation:Decade Resistance Box, 4