Vornado™ Mini Vortexer
Vornado™ Mini Vortexer
Supplier:   Ward's
Vornado™ Mini Vortexer
Supplier:  Ward's
Certificates
Vornado™ Mini Vortexer
3 Options Available Below
Product Details & Documents
Powerful vortexing for tubes up to 50 ml.
  • Insta-TouchTM pressure activated operation
  • "Mini" sized, less than 4" wide
  • 2 year warranty
Powerful is the best word to describe this mini vortexer. With a 4 mm orbit and fixed speed of 2800 rpm, the Vornado instantly vortexes even the largest samples, included nearly full 50 ml tubes. Despite its powerful motor, the Vornado has a footprint of less than 4 x 4 inches, allowing it to fit on even the most crowded bench.
Insta-touch operation means that the motor is automatically activated with slight pressure on the center cup head. Operation is smooth, quiet and efficient. When pressure is removed, operation immediately ceases. The Vornado's unique head design prevents liquids from entering the housing, prolonging motor life.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 470230-478
  • 470230-480
  • 470230-482
  • Orbit:
  • 4 mm
  • 4 mm
  • 4 mm
  • Electrical:
  • 115 V, 50/60 Hz, 0.7 A
  • 115 V, 50/60 Hz, 0.7 A
  • 115 V, 50/60 Hz, 0.7 A
  • Dimensions:
  • 3.7 x 3.9 x 2.6"
  • 3.7 x 3.9 x 2.6"
  • 3.7 x 3.9 x 2.6"
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Electrical
    Speed
    Availability
    Price
  • Vornado™ Mini Vortexer, Grey Cup Head
    115 V, 50/60 Hz, 0.7 A
    2800 rpm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $343.83
    $343.83
    Each
       $343.83
    Supplier Number: BV101_SE
    Spécifications:
    More
    Specifications
    Cat. No.
    470230-478
    Orbit
    4 mm
    Electrical
    115 V, 50/60 Hz, 0.7 A
    Dimensions
    3.7 x 3.9 x 2.6"
  • Vornado™ Mini Vortexer, Blue Cup Head
    115 V, 50/60 Hz, 0.7 A
    2800 rpm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $390.90
    $390.90
    Each
       $390.90
    Supplier Number: BV101-B
    Spécifications:
    More
    Specifications
    Cat. No.
    470230-480
    Orbit
    4 mm
    Electrical
    115 V, 50/60 Hz, 0.7 A
    Dimensions
    3.7 x 3.9 x 2.6"
  • Vornado™ Mini Vortexer, Green cup head
    115 V, 50/60 Hz, 0.7 A
    2800 rpm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $390.90
    $390.90
    Each
       $390.90
    Supplier Number: BV101-G_SE
    Spécifications:
    More
    Specifications
    Cat. No.
    470230-482
    Orbit
    4 mm
    Electrical
    115 V, 50/60 Hz, 0.7 A
    Dimensions
    3.7 x 3.9 x 2.6"