Battery Holder | Avantor
Battery Holder
Battery Holder
Catalog # 470230-352
Supplier:   Wards
Fournisseur #:  PH0957D
Battery Holder
Catalog # 470230-352
Supplier:  Wards
Battery Holder
Battery Holder
Battery Holder
Price
$64.80
Log in pour la disponibilité
Each
Product Details & Documents
Ideal for studying electricity.
  • Allows you to connect up to 4 D cell batteries in series or parallel (comes configured in series, can be converted though - may require additional wires)
  • Perfect for electronics, physics, or even biology experiments
  • Batteries and cables not included
  • Variable battery power supply
The variable power supply that hold up to 4 D cells is a great tool for use when studying electricity (Batteries and cables not included).
Spécifications
  • Désignation:
    Battery Holder