Plastic Pulley Triple | Avantor
Plastic Pulley Triple
Plastic Pulley, Triple
Catalog # 470230-092
Supplier:   Wards
Fournisseur #:  PH0281C
Plastic Pulley Triple
Catalog # 470230-092
Supplier:  Wards
Plastic Pulley Triple
Plastic Pulley Triple
Plastic Pulley, Triple
Liquidation
$12.45
Vente
Valable jusqu'au 31/12/2026
Log in pour la disponibilité
Each
Product Details & Documents
Economical pulley for simple machine experiments.
  • Triple plastic pulley, in tandem
  • Pully diameters of 25, 38 and 50 mm
  • Low friction design
  • Open sided frame for ease of use
A high quality, triple plastic pulley in tandem, on an open sided steel frame, with hooks at each end carrying three ball bearing plastic pulleys with diameters of 25, 38 and 50 mm respectively. The open sided frame greatly facilitates stringing up as no 'threading' is involved. Inner gap of pulley measures 3 mm, overall length is 17.6 cm. Capable of holding over 30 lbs of weight.
Spécifications
  • Désignation:
    Plastic Pulley, Triple