Metal Pulley Double Parallel
Catalogue # 470230-076
Fournisseur: Wards
Spécifications
- Description:Metal Pulley, Double Parallel
- Cat. no.:470230-076
Spécifications
À propos de cet article
Precisely centered aluminum pulleys.
- Aluminum pulleys
- 5cm dia.
- Durable aluminum construction
- Precisely centered
Durable aluminum pulleys with 5cm diameters, mounted in a metal frame.