ContactUsLinkComponent
Metal Pulley Double Parallel
Catalogue # 470230-076
Fournisseur:  Wards
undefined
Metal Pulley Double Parallel
Catalogue # 470230-076
Fournisseur:  Wards
Supplier Number:  PH0278C
Spécifications
  • Description:
    Metal Pulley, Double Parallel
  • Cat. no.:
    470230-076
Spécifications
À propos de cet article
Precisely centered aluminum pulleys.
  • Aluminum pulleys
  • 5cm dia.
  • Durable aluminum construction
  • Precisely centered
Durable aluminum pulleys with 5cm diameters, mounted in a metal frame.