Friction Block Felt with Hook | Avantor
Friction Block Felt with Hook
Friction Block Felt with Hook
Catalog # 470230-070
Supplier:   Wards
Fournisseur #:  PH0201605F
Friction Block Felt with Hook
Catalog # 470230-070
Supplier:  Wards
Friction Block Felt with Hook
Friction Block Felt with Hook
Friction Block Felt with Hook
Price
$26.70
Log in pour la disponibilité
Each
Product Details & Documents
Friction blocks are useful for teaching the basics of force and motion.
  • Wooden friction block with c-hook attached at one end
  • Use with a spring scale to measure force and mass - all the numbers you need to solve a variety of motion and force problems
  • Differing pine and felt surfaces allow for comparative study
  • Hands-on, economic tool for teaching fundamental physics concepts to students of any age
  • Manufactured in the US
Friction blocks are useful for teaching the basics of force and motion in a simple, yet effective way. Fitted with a c-hook, the block can be attached to a spring scale to measure incident forces. Slide the block along a flat surface, or pull it up an incline. Observe the block by itself, or add weights to the top. Measure differences in coefficient of friction between the felt and pine surfaces. A friction block used in tandem with a spring scale is a powerful tool for teaching students about friction, kinetic and potential energy, acceleration, and force. This block is sourced and manufactured in the United States.
Spécifications
  • Désignation:
    Friction Block Felt with Hook