Spouting Cylinder
Spouting Cylinder
Catalog # 470230-064
Supplier: Wards
Supplier #: PH0196
Jump to:
Product Details & Documents
Durable shatter-proof plastic.
- Demonstrating principles of Snell's Law
- 2.3" (60 mm) dia. pipe
- 15.5" (400 mm) total height
Demonstrate Snell's Law from an acrylic pipe. The pipe is 15.5" tall and 2.3" in diameter and has holes at predetermined heights.
Specifications
- Désignation:Spouting Cylinder