Specific Gravity Cylinders
Specific Gravity Cylinders
Catalog # 470230-040
Supplier: Wards
Fournisseur #: PH0110
Jump to:
Product Details & Documents
Four different metal cylinders each measuring 38 mm long x 10 mm in diameter.
- 4 metal cylinders for the study of gravity, density, and mass
- Aluminum, zinc, copper and steel
- Comes with hook holes and removable hooks to be used in a variety of experiments
Four different metal cylinders each measuring 38 mm long x 10 mm in diameter, made of aluminum, zinc, copper and steel. Comes with hook holes and removable hooks to be used in a variety of experiments.
Spécifications
- Désignation:Specific Gravity Cylinders