ContactUsLinkComponent
Monkey Hunter Demo
Catalog # 470230-008
Supplier:  Wards
undefined
Monkey Hunter Demo
Catalog # 470230-008
Supplier:  Wards
Supplier Number:  MNKHT
Specifications
  • Description:
    Monkey Hunter Demonstration
  • Cat. no.:
    470230-008
Specifications
About this item
This kit is used to perform the classic experiment of the 'Monkey and the Hunter'.
  • Perform a visual representation of horizontal and vertical motion
  • System can be applied to all age levels as an introduction to projectile theory
  • Kit operates using a low voltage power supply at about 4V (not included)
  • Kit contains detailed operating instructions and discussion material
  • Please read instructions carefully before operating
This kit is used to perform the classic experiment of the 'Monkey and the Hunter'. It is a great demonstration of two-dimensional motion concepts and the independence of the horizontal and vertical motion of a projectile. The kit contains a launcher, a projectile, connecting cables, a stuffed monkey and an electromagnet. You will need a low voltage power supply that operates at 4V d.c. The current requirement is about 0.4A. The monkey is suspended from the electromagnet by a metal cylinder on its head and when the launcher is triggered the electromagnet will release the monkey. The student can then apply their observations to study formulas related to the experiment. The kit contains a detailed instruction manual.