Demo Relay Switch
Demo Relay Switch
470229-994
: Wards
: DER14
Ideal tool for studying and observing the basic concepts of a relay.
- Relay demonstration device
- Circuit Board with clear labels
- Displayed on sturdy base
- Circuit powering the coil is completely separate from the circuit switched on by the relay
The relay demonstration is an ideal tool for studying and observing the basic concepts of a relay (an electrically operated switch that opens and closes circuits) in a visually and kinesthetically effective manner.
- Désignation:Demo Relay Switch