Standard Methods Agar Prepared Bottled Media, BD Diagnostics
Fournisseur: BD
Certificats
À propos de cet article
BD Prepared Bottled Media are ready-to-use formulations intended for the detection of micororganisms from pharmaceutical, food, environmental or water testing samples. They are offered in a wide variety of bottle styles and fill volumes designed to meet customer requirements.
- Use of highest quality BBL and Difco brand dehydrated culture media for preparation
- Conformation to current USP and EP requirements (see package labeling for details)
- Color-coded caps and/or labeling for most products, providing easy product identification
- Transparent labeling to allow a clear view of media and culture
Septum/Screw Cap style has a black, phenolic screw cap with septum.
Screw Cap is the same as Septum/Screw Cap but without septum.
Frequently Bought Together