VWR® Erlenmeyer Flask with Standard Taper Outer Joint
Fournisseur: VWR International
Certificats
À propos de cet article
High-quality durable flask featuring a ground joint and uniform wall thickness and are a great value.
- Manufactured from ASTM E438 Type 1
- Class A borosilicate 3.3 expansion glass
VWR® Erlenmeyer flasks are used to measure, mix or hold fluids or chemicals commonly used in laboratories.
Frequently Bought Together