ContactUsLinkComponent
Nutrient Agar Slants, Prepared Media in Tubes, BD Diagnostics
Catalogue # CA-90001-172
Fournisseur:  BD
Numéro CAS:  
Nutrient Agar Slants, Prepared Media in Tubes, BD Diagnostics
Catalogue # CA-90001-172
Fournisseur:  BD
Supplier Number:  220971
Numéro CAS:  
Spécifications
  • Pack type:
    Tube
  • Description:
    Nutrient Agar Slants
  • Cat. no.:
    CA-90001-172
Spécifications
À propos de cet article
Media for the isolation and identification of microorganisms.
  • Ideal for cultures requiring prolonged incubation
  • Made of borosilicate glass
  • Securely fitted screw caps to prevent loss of moisture
Media for the isolation and identification of microorganisms. Tubed media are ideal for cultures requiring prolonged incubation, since the caps can be closed tightly to prevent dehydration and subsequent inhibition of growth. Long-necked, rubber-lined screw caps eliminate danger of contamination. Larger-sized tubes with wider diameters permit easy inoculation and handling of media. Tubes are made of borosilicate glass and have securely fitted screw caps to prevent loss of moisture. Caps contain a resilient rubber liner that resists displacement even during autoclaving.