Middlebrook and Cohn 7H10 Agar Slants, Prepared Media in Tubes, BD Diagnostics
Catalog # CA-90001-164
Supplier: BD
CAS Number:
Specifications
- Pack type:Tube
- Description:Middlebrook and Cohn 7H10 Agar Slants
- Cat. no.:CA-90001-164
- Tube Size:20×148 mm
- Supplier No.:220959
Specifications
About this item
Media for the isolation and identification of microorganisms.
- Ideal for cultures requiring prolonged incubation
- Made of borosilicate glass
- Securely fitted screw caps to prevent loss of moisture
Media for the isolation and identification of microorganisms. Tubed media are ideal for cultures requiring prolonged incubation, since the caps can be closed tightly to prevent dehydration and subsequent inhibition of growth. Long-necked, rubber-lined screw caps eliminate danger of contamination. Larger-sized tubes with wider diameters permit easy inoculation and handling of media. Tubes are made of borosilicate glass and have securely fitted screw caps to prevent loss of moisture. Caps contain a resilient rubber liner that resists displacement even during autoclaving.