Scandium Standards, CRM for AA and ICP, Assurance Grade, SPEX CertiPrep®
Catalogue # CA11023-684
Fournisseur: Spex Certiprep
Numéro CAS:
Spécifications
- Concentration:1000 µg/ml
- Matrix:2% HNO₃
- Pack type:Plastic Bottle
- Size:125 ml
- Cat. no.:CA11023-684
- Element:Scandium
- Supplier No.:PLSC2-2Y
Spécifications
À propos de cet article
- Traceable to NIST
- Uncertainty of standards is reported on the Certificate of Analysis
- Manufactured by experienced chemists to ensure highest quality standards
Caution: 1000 µg/mL = 1000 ppm