Ward's® Essentials Spacesuit Design Challenge
Ward's® Essentials Spacesuit Design Challenge
Spacesuit Design Challenge, 10
Catalog # 470231-276
Supplier:   Science First
Supplier #:  650-1000
Ward's® Essentials Spacesuit Design Challenge
Catalog # 470231-276
Supplier:  Science First
Ward's® Essentials Spacesuit Design Challenge
Ward's® Essentials Spacesuit Design Challenge
Spacesuit Design Challenge, 10
Price
$80.99
Log in for availability
Each Kit of 1
Product Details & Documents
The Perfect Introduction to Temperature and Conduction.
  • Encourages teamwork
    Students apply standard engineering design processes while applying economics
    Investigation includes teaching guides
    Includes various materials to simulate spacesuit construction
    Aligned with NGSS MS-PS3-3, MS-ETS1-1, MS-ETS1-2, MS-ETS1-3, MS-ETS1-4
Students are presented with a defined set of materials and asked to design a spacesuit that can help maintain the body temperature of an astronaut in the extreme heat and cold of outer space. Water-filled 'Space Worms' are used as test subjects that mimic the water composition of the adult human body. Students must create a budget and plan their testing rounds accordingly. Students collect data on internal temperature changes of the Space Worms and evaluate which suits work best given the economic constraints. This kit includes enough materials for 10 student groups.
Ordering information: Kit includes water-filled Space Worms, a variety of insulating materials, thermometers, and teacher’s guide.
Specifications
  • Désignation:
    Spacesuit Design Challenge
  • No. of Lab Groups:
    10