ContactUsLinkComponent
Fractional Distillation of Crude Oil
Fractional Distillation of Crude Oil
Catalog # 470225-448
Supplier:  Ward's Science
undefined
Fractional Distillation of Crude Oil
Catalog # 470225-448
Supplier:  Ward's Science
Supplier Number:  AD-KK7107-KIT
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Fractional Distillation of Crude Oil
  • No. of Lab Groups:
    15
  • Cat. no.:
    470225-448
  • Supplier No.:
    AD-KK7107-KIT
Specifications
About this item
Understand Distillation and Intermolecular Forces with this Real Life Application.
  • Hands on lab activity that illustrates real life process
  • Instructions and materials for 15 lab groups
Crude oil is a complex mixture of many useful hydrocarbons, such as gasoline, kerosene, butane, and motor oil. In order to be of use, these hydrocarbons must first be separated from each other. The process of separation, known as fractional distillation, takes advantage of the fact that these components have varying boiling and condensation points. In this activity, students will complete a fractional distillation experiment to simulate the fractionation of crude oil. As part of this activity students will understand how a difference in boiling points can be utilized to separate a solution with three different components and how intramolecular forces can affect the boiling point of a substance.