ContactUsLinkComponent
Safety Storage Container, Stainless Steel, Bürkle
Catalog # 76102-202
Supplier:  BURKLE INC
undefined
Safety Storage Container, Stainless Steel, Bürkle
Catalog # 76102-202
Supplier:  BURKLE INC
Supplier Number:  2601-4010
Specifications
  • Capacity:
    10 L (2.64 gal.)
  • Description:
    Safety Storage/Transport Container with tab and pressure relief valve
  • Material:
    Stainless steel
  • Cat. no.:
    76102-202
  • O.D.×H:
    260×330 mm (12.9×10.24")
Specifications
About this item
Safety containers for transport and storage are made of high-quality stainless steel AISI 304 (1.4301)/AISI 316 Ti (1.4571). Containers have a uniform wall thickness of 0.8 mm (0.03").
  • Provide a high degree of chemical resistance
  • Excess pressure valve allows pressure balance to occur at approximately 0.3 bar (4.35 psi)
  • Self-closing tap provides constant ventilation that can be set via a plastic lever on the screw cap
  • Perfluoroalkoxy copolymers (PFA) window enables a visual contents check when filling and emptying
Containers provide protection from explosion or flames through an excess pressure valve, along with seals made of PTFE.
Thread: 1½"
Thread (outflowing side): ¾"
Certifications: Accredited for UN-transports according to ADR, RID,IMDG Code, ICAO-TI, GGVSEB, GGVSee.
Packaging: One container per package.