ZR-96 MagStand, Zymo Research
Catalog # 76005-120
Supplier: Zymo Research
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:ZR-96 MagStand
- Size:96 well
- Cat. no.:76005-120
Specifications
About this item
The ZR-96 MagStand is a magnetic stand for the separation of magnetic beads in standard 96-well format plates, especially for EZ-96 DNA Methylation MagPrep product line, ZR-96 Genomic DNA MagPrep, Zyppy-96 Plasmid MagBead Miniprep, and Direct-zol-96 MagBead RNA.