KALON Glass Dressing Jars, Profex
36311-052
Small Business Enterprise
- Ø:7.6 cm (3")
- Color:Clear
- Description:Glass Dressing Jar with Sandblast Area for Writing
- Height:7.6 cm (3")
- Cat. no.:36311-052
- Supplier No.:1174SSA
Made of clear heavy glass, with stainless steel covers.