Mueller Hinton Broth, Hardy Diagnostics
10324-388
Product available on GSA Advantage®
:
- Pack type:Tube
- Description:Mueller Hinton Broth
- Cat. no.:10324-388
- Tube Size:13×100 mm
- Supplier No.:R38
For the preparation of suspensions for disk diffusion sensitivity testing.