Biox Multi-Well Culture Plates
Supplier: Avantor
Certificates
About this item
Cell Culture plates are sterile, constructed of prime virgin polystyrene, and a great value.
- Prime virgin polysyrene
- Individually wrapped packaging
- Sterile
- Cover design minimizes cross contamination
- Thin bottom wall increases thermal consisency
- Flat well bottom provides excellent clarity
TC treated plates come with a cover and feature flat bottoms for excellent clarity for microsopic examination of samples.
Certifications: ISO Certified: GB/T 19001-2008/ISO 9001:2008.
Frequently Bought Together