Biox Culture Dishes
Supplier: TWD Scientific
Certificates
About this item
Sterile and constructed of optically clear material, these cell culture dishes offer high quality at a great price.
- High-quality medical grade polystyrene
- Vacuum plasma surface treatment ensures excellent cell adhesion
- Lid design promotes effective gas exchange
- External grip for easier handling (available on 2 models)
- Electron beam sterilized
- Non-pyrogenic and non-cytotoxic
- Dnase- & Rnase-free, human DNA-free
These TC treated culture dishes have vented lids and raised edges for convenient stacking.
Certifications: ISO Certified: GB/T 19001-2008/ISO 9001:2008.
Frequently Bought Together