Biox Tissue Culture Flasks
High quality polystyrene cell culture flasks are sterile, stackable, and a great value.
- High-quality medical grade polystyrene
- Plug caps completely isolate the culture from the outside environment
- Filter Caps prevent cross-contamination while meeting the requirements for gas exchange
- Electron beam sterilized
- Short, wide, angled neck design for easy access
- Non-pyrogenic and non-cytotoxic
- DNase- & RNase-free, Human DNA-free
TC treated cell culture flasks with plug or filter caps allow for complete isolation from the outside environment and prevent cross contamination while allowing for gas exchange.
: ISO Certified: GB/T 19001-2008/ISO 9001:2008.
High quality polystyrene cell culture flasks are sterile, stackable, and a great value.
- High-quality medical grade polystyrene
- Plug caps completely isolate the culture from the outside environment
- Filter Caps prevent cross-contamination while meeting the requirements for gas exchange
- Electron beam sterilized
- Short, wide, angled neck design for easy access
- Non-pyrogenic and non-cytotoxic
- DNase- & RNase-free, Human DNA-free
TC treated cell culture flasks with plug or filter caps allow for complete isolation from the outside environment and prevent cross contamination while allowing for gas exchange.
: ISO Certified: GB/T 19001-2008/ISO 9001:2008.
Product Family Options
- DésignationTotal VolumeWorking VolumeGrowth AreaCap TypeSterility
- Clear Polystyrene (TC-Treated) Cell Culture Flasks with Filter Cap Assembled60 ml7 ml25 cm2Filter capSterile-: 07-8025
- Clear Polystyrene (TC-Treated) Cell Culture Flasks with Filter Cap Assembled250 ml25 ml75 cm2Filter capSterile-: -
- Clear Polystyrene (TC-Treated) Cell Culture Flasks with Plug Cap Assembled60 ml7 ml25 cm2Plugged capSterileCase of 200$486.40: BX07-9025
- Clear Polystyrene (TC-Treated) Cell Culture Flasks with Plug Cap Assembled250 ml25 ml75 cm2Plugged capSterile-: BX07-9075