Thermal Printer, Ohaus | Avantor
Thermal Printer, Ohaus
Thermal Printer, 120 - 230 VAC, Thermal paper
Catalog # 97009-692
Supplier:   Ohaus
Fournisseur #:  80251992
Thermal Printer, Ohaus
Catalog # 97009-692
Supplier:  Ohaus
Thermal Printer, Ohaus
Thermal Printer, 120 - 230 VAC, Thermal paper
Price
$1,821.24
Log in pour la disponibilité
Each
Product Details & Documents
This thermal printer has a small footprint
  • Compact and lightweight
  • Easy paper loading
  • Paper near end detection
  • Included AC power adapter
It is user-friendly and offers unsurpassed reliability.
Certifications: CE Marking, TUV-GS, FCC Class B, VCCI Class B (AC adapter PSE / UL /C-UL / TUV-GS)
Spécifications
  • Désignation:
    Thermal Printer
  • Electrical:
    120 - 230 VAC
  • Paper Type:
    Thermal paper
  • Cat. No.:
    97009-692
  • Length:
    191 mm (7.5")
  • Height:
    90 mm (3.5")
  • Width:
    100 mm (3.9")