Magne-Box™ Magnetic Polycarbonate Cryoboxes, Argos Technologies
Fournisseur: Antyila Scientific
Certificats
À propos de cet article
Magne-Box™ freezer boxes are the first of its kind to offer a magnetic lid closure.
- Boxes fit in standard size freezer racks
- Polycarbonate design withstands temperatures from –196 to 121 °C
- Slot openings on the bottom of boxes allow for drainage
- 25-, 81-, and 100-Place boxes accommodate 2 mL cryovials
- 81-Place Tall boxes accommodate 4 to 5 mL cryovials
Each box features magnets that securely connect the base to the hinged lid. This allows one-handed retrieval from freezer racks and ensures that samples stay safe. Polycarbonate boxes have clear lids with colored inserts and numbered grids to facilitate easy sample identification. Available with 25-, 81-, and 100-place divisions.
Frequently Bought Together