Magnetic Bar Magnets
Magnetic Bar Magnets
Catalog # 470229-308
Supplier: United Scientific Supplies
Fournisseur #: MPC080
Jump to:
Product Details & Documents
Plastic coated bar magnets.
- Color-coded ends
- Strong Alnico magnets
These alnico magnets are encased in red and blue plastic to provide extra durability. Each magnet measures 80x15x10mm. Shipped as a pair.
Spécifications
- Désignation:Magnetic Bar Magnets