Engineering Hydraulic Syringes
Supplier:   Avantor
Engineering Hydraulic Syringes
Supplier:  Avantor
Certificates
Engineering Hydraulic Syringes
Engineering Hydraulic Syringes
2 Options Available Below
Product Details & Documents
Perfect for Makerspace and hydraulic projects.
Convenient packs of 10 for use in a variety of Makerspace and engineering challenges. All come with a lure lock fitting that fits snuggly into vinyl tubing, but can not be fitted with a needle.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • 30 mL Superlock Syringe
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $37.79
    $37.79
    Pack of 10
       $37.79
  • 60 mL Superlock Syringe
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $55.99
    $55.99
    Pack of 10
       $55.99